5-Methylpyridazin-3(2H)-one ≥99%

Supplier: Matrix Scientific

070470-1G 070470-5G
101944-938EA 40 USD
101944-938 101944-940
5-Methylpyridazin-3(2H)-one ≥99%
5-Methylpyridazin-3(2H)-one

Hazard: Irritant

Formula: C5H6N2O
MDL Number: MFCD09743644
CAS Number: 54709-94-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR