5-Bromo-1,4-dimethyl-1H-indazole ≥97%

Supplier: Matrix Scientific

070466-1G 070466-5G
101944-924EA 1221.54 USD
101944-924 101944-926
5-Bromo-1,4-dimethyl-1H-indazole ≥97%
5-Bromo-1,4-dimethyl-1H-indazole

Hazard: Irritant

Formula: C₉H₉BrN₂
MW: 225.09 g/mol
MDL Number: MFCD12028624
CAS Number: 1159511-80-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR