4-Bromo-1,5-dimethyl-1H-indazole ≥97%

Supplier: Matrix Scientific

070463-1G 070463-5G
101944-910EA 1221.54 USD
101944-910 101944-912
4-Bromo-1,5-dimethyl-1H-indazole ≥97%
4-Bromo-1,5-dimethyl-1H-indazole

Hazard: Irritant

Formula: C₉H₉BrN₂
MDL Number: MFCD12028620
CAS Number: 1159511-77-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR