2,3-Dihydro-1H-indole-7-carbonitrile ≥96%

Supplier: Matrix Scientific

070462-1G 070462-5G 070462-25G
101944-904EA 640 USD
101944-904 101944-906 101944-908
2,3-Dihydro-1H-indole-7-carbonitrile ≥96%
2,3-Dihydro-1H-indole-7-carbonitrile

Hazard: Irritant

Formula: C₉H₈N₂
MDL Number: MFCD01830669
CAS Number: 115661-82-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR