4-(Bromomethyl)-1-methyl-1H-indazole ≥95%

Supplier: Matrix Scientific

070461-1G 070461-5G
101944-900EA 1221.54 USD
101944-900 101944-902
4-(Bromomethyl)-1-methyl-1H-indazole ≥95%
4-(Bromomethyl)-1-methyl-1H-indazole

Hazard: Irritant

Formula: C₉H₉BrN₂
MDL Number: MFCD11052619
CAS Number: 1092961-03-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR