2-Methyl-5-(tributylstannyl)thiazole ≥96%

Supplier: Matrix Scientific

070458-500MG 070458-1G 070458-5G
101944-884EA 489.23 USD
101944-884 101944-886 101944-888
2-Methyl-5-(tributylstannyl)thiazole ≥96%
2-Methyl-5-(tributylstannyl)thiazole

Hazard: Irritant

Formula: C₁₆H₃₁NSSn
MDL Number: MFCD01319032
CAS Number: 223418-75-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR