2-(Methylsulfonyl)-5-(tributylstannyl)thiazole ≥94%

Supplier: Matrix Scientific

070457-500MG 070457-1G 070457-5G
101944-878EA 1135.38 USD
101944-878 101944-880 101944-882
2-(Methylsulfonyl)-5-(tributylstannyl)thiazole ≥94%
2-(Methylsulphonyl)-5-(tributylstannyl)thiazole

Hazard: Irritant

Formula: C₁₆H₃₁NO₂S₂Sn
MDL Number: MFCD01319035
CAS Number: 1245816-12-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR