2-Chloro-4-(tributylstannyl)thiazole ≥90%
Supplier: Matrix Scientific
070454-500MG
070454-1G
070454-5G
101944-860EA
1338.46
USD
101944-860
101944-862
101944-864
2-Chloro-4-(tributylstannyl)thiazole ≥90%
2-Chloro-4-(tributylstannyl)thiazole
Hazard: Irritant
Formula:
C₁₅H₂₈ClNSSn |
MDL Number:
MFCD16140394 CAS Number: 1245816-11-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...