2-(Methylthio)-4-(tributylstannyl)thiazole ≥94%

Supplier: Matrix Scientific

070453-500MG 070453-1G 070453-5G
101944-854EA 1338.46 USD
101944-854 101944-856 101944-858
2-(Methylthio)-4-(tributylstannyl)thiazole ≥94%
2-(Methylthio)-4-(tributylstannyl)thiazole

Hazard: Irritant

Formula: C₁₆H₃₁NS₂Sn
MDL Number: MFCD16140393
CAS Number: 446286-06-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR