2-(Methylsulfonyl)-4-(tributylstannyl)thiazole ≥94%
Supplier: Matrix Scientific
070452-500MG
070452-1G
070452-5G
101944-848EA
1135.38
USD
101944-848
101944-850
101944-852
2-(Methylsulfonyl)-4-(tributylstannyl)thiazole ≥94%
2-(Methylsulfonyl)-4-(tributylstannyl)thiazole
Hazard: Irritant
Formula:
C₁₆H₃₁NO₂S₂Sn |
MDL Number:
MFCD17015019 CAS Number: 1245816-14-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...