3-(4-(Tributylstannyl)thiazol-2-yl)oxetan-3-ol

Supplier: Matrix Scientific

070451-500MG 070451-1G 070451-5G
101944-842EA 1687.69 USD
101944-842 101944-844 101944-846
3-(4-(Tributylstannyl)thiazol-2-yl)oxetan-3-ol
3-(4-(Tributylstannyl)thiazol-2-yl)oxetan-3-ol

Hazard: Irritant

Formula: C₁₈H₃₃NO₂SSn
MDL Number: MFCD16140389
CAS Number: 1245816-13-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR