2-(4-(Tributylstannyl)thiazol-2-yl)-2-propanol ≥95%

Supplier: Matrix Scientific

070450-500MG 070450-1G 070450-5G
101944-836EA 989.23 USD
101944-836 101944-838 101944-840
2-(4-(Tributylstannyl)thiazol-2-yl)-2-propanol ≥95%
2-(4-(Tributylstannyl)thiazol-2-yl)-2-propanol

Hazard: Irritant

Formula: C₁₈H₃₅NOSSn
MDL Number: MFCD16140388
CAS Number: 1245816-16-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR