5-Bromo-4-(tributylstannyl)pyrimidine ≥94%

Supplier: Matrix Scientific

070448-500MG 070448-1G 070448-5G
101944-824EA 1396.92 USD
101944-824 101944-826 101944-828
5-Bromo-4-(tributylstannyl)pyrimidine ≥94%
5-Bromo-4-(tributylstannyl)pyrimidine

Hazard: Irritant

Formula: C₁₆H₂₉BrN₂Sn
MDL Number: MFCD16170450
CAS Number: 1416437-21-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR