2-Chloro-4-(tributylstannyl)pyrimidine ≥93%

Supplier: Matrix Scientific

070447-1G 070447-5G
101944-820EA 900 USD
101944-820 101944-822
2-Chloro-4-(tributylstannyl)pyrimidine ≥93%
2-Chloro-4-(tributylstannyl)pyrimidine

Hazard: Irritant

Formula: C₁₆H₂₉ClN₂Sn
MDL Number: MFCD16170326
CAS Number: 446286-25-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR