3-Fluorodihydro-2H-pyran-4(3H)-one ≥94%

Supplier: Matrix Scientific

Synonyms: 3-Fluorooxan-4-one

070446-250MG 070446-500MG 070446-1G
101944-812EA 407.69 USD
101944-812 101944-814 101944-816
3-Fluorodihydro-2H-pyran-4(3H)-one ≥94%
3-Fluorodihydro-2H-pyran-4(3H)-one

Hazard: Irritant

Formula: C₅H₇FO₂
MW: 118.11 g/mol
MDL Number: MFCD11977351
CAS Number: 624734-19-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR