tert-Butyl 3,3-difluoro-4-oxopiperidine-1-carboxylate hydrate ≥97%

Supplier: Matrix Scientific

070445-500MG 070445-1G 070445-5G
101944-806EA 150.77 USD
101944-806 101944-808 101944-810
tert-Butyl 3,3-difluoro-4-oxopiperidine-1-carboxylate hydrate ≥97%
tert-Butyl 3,3-difluoro-4-oxopiperidine-1-carboxylate hydrate

Hazard: Irritant

Formula: C₁₀H₁₅F₂NO₃·H₂O
MW: 253.243 g/mol
MDL Number: MFCD19443979
CAS Number: 1400264-85-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR