3-Trifluoromethylpyrrolidine hydrochloride ≥97%

Supplier: Matrix Scientific

Synonyms: 3-(Trifluoromethyl)pyrrolidine hydrochloride

070439-500MG 070439-1G 070439-5G
101944-772EA 524.62 USD
101944-772 101944-774 101944-776
3-Trifluoromethylpyrrolidine hydrochloride ≥97%
3-Trifluoromethylpyrrolidine hydrochloride

Hazard: Irritant

Formula: C₅H₉ClF₃N
MW: 175.58 g/mol
MDL Number: MFCD03452798
CAS Number: 1189485-03-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR