3-Oxocyclobutyl benzoate ≥97%

Supplier: Matrix Scientific

070438-500MG 070438-1G 070438-5G
101944-766EA 538.46 USD
101944-766 101944-768 101944-770
3-Oxocyclobutyl benzoate ≥97%
3-Oxocyclobutyl benzoate

Hazard: Irritant

Formula: C₁₁H₁₀O₃
MDL Number: MFCD16140376
CAS Number: 1081559-36-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR