1-(tert-Butoxycarbonylamino)-2,2-difluorocyclopropanecarboxylic acid ≥97%

Supplier: Matrix Scientific

070437-250MG 070437-500MG 070437-1G
101944-760EA 1920 USD
101944-760 101944-762 101944-764
1-(tert-Butoxycarbonylamino)-2,2-difluorocyclopropanecarboxylic acid ≥97%
1-(tert-Butoxycarbonylamino)-2,2-difluorocyclopropanecarboxylic acid

Hazard: Irritant

Formula: C₉H₁₃F₂NO₄
MDL Number: MFCD16039589
CAS Number: 796882-45-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR