1-Amino-1-cyclopropanecarbonitrile hydrochloride ≥97%

Supplier: Matrix Scientific

070436-5G 070436-25G
101944-756EA 60 USD
101944-756 101944-758
1-Amino-1-cyclopropanecarbonitrile hydrochloride ≥97%
1-Amino-1-cyclopropanecarbonitrile hydrochloride

Hazard: Toxic

Formula: C₄H₇ClN₂
Melting Pt: ∼ 220 °C (dec.)
MDL Number: MFCD04114063
CAS Number: 127946-77-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR