6-Chloropyrazolo[1,5-a]pyridine-2-carboxylic acid methyl ester ≥97%

Supplier: Matrix Scientific

070435-1G 070435-5G
101944-750EA 1396.92 USD
101944-750 101944-752
6-Chloropyrazolo[1,5-a]pyridine-2-carboxylic acid methyl ester ≥97%
6-Chloropyrazolo[1,5-a]pyridine-2-carboxylic acid methyl ester

Hazard: Irritant

Formula: C₉H₇ClN₂O₂
MW: 210.62 g/mol
MDL Number: MFCD16140374
CAS Number: 1222709-26-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR