3,3-Difluorocyclobutyl benzoate ≥97%

Supplier: Matrix Scientific

070433-250MG 070433-500MG 070433-1G
101944-738EA 1920 USD
101944-738 101944-740 101944-742
3,3-Difluorocyclobutyl benzoate ≥97%
3,3-Difluorocyclobutyl benzoate

Hazard: Irritant

Formula: C₁₁H₁₀F₂O₂
MDL Number: MFCD16039588
CAS Number: 1215071-19-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR