6,6-Difluoro-3-azabicyclo[3.1.0]hexane hydrochloride ≥97%

Supplier: Matrix Scientific

070432-100MG 070432-250MG 070432-500MG
101944-732EA 737.35 USD
101944-732 101944-734 101944-736
6,6-Difluoro-3-azabicyclo[3.1.0]hexane hydrochloride ≥97%
6,6-Difluoro-3-azabicyclo[3.1.0]hexane hydrochloride

Hazard: Irritant

Formula: C₅H₈ClF₂N
MW: 155.58 g/mol
MDL Number: MFCD16039587
CAS Number: 1215071-13-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR