2-Amino-2-(3,3-difluorocyclobutyl)acetonitrile hydrochloride

Supplier: Matrix Scientific

070430-250MG 070430-500MG 070430-1G
101944-720EA 1920 USD
101944-720 101944-722 101944-724
2-Amino-2-(3,3-difluorocyclobutyl)acetonitrile hydrochloride
2-Amino-2-(3,3-difluorocyclobutyl)acetonitrile hydrochloride

Hazard: Irritant

Formula: C₆H₉ClF₂N₂
MDL Number: MFCD16039585
CAS Number: 1215166-79-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR