4-Bromo-2-fluorophenylboronic acid ≥98%
Supplier: Matrix Scientific
Synonyms:
4-Bromo-2-fluorobenzeneboronic acid
069396-1G
069396-5G
069396-25G
101939-930EA
175.38
USD
101939-930
101939-932
101939-934
4-Bromo-2-fluorophenylboronic acid ≥98%
4-Bromo-2-fluorophenylboronic acid
Hazard: Irritant
Formula:
C₆H₅BBrFO₂ MW: 218.82 g/mol |
MDL Number:
MFCD01318110 CAS Number: 216393-64-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...