2-Oxo-7-azaspiro[3.5]nonane-7-carboxylate-tert-butyl ester ≥95%

Supplier: Matrix Scientific

Synonyms: tert-Butyl 2-oxo-7-azaspiro[3.5]nonane-7-carboxylate, 7-Azaspiro[3.5]nonan-2-one, N-BOC protected, 2-Oxo-7-azaspiro[3.5]nonane-7-carboxylate tert-butyl ester

069394-250MG 069394-1G
101939-920EA 295.54 USD
101939-920 102551-748
2-Oxo-7-azaspiro[3.5]nonane-7-carboxylate-tert-butyl ester ≥95%
2-Oxo-7-azaspiro[3.5]nonane-7-carboxylate-tert-butyl ester

Hazard: Irritant

Formula: C₁₃H₂₁NO₃
MW: 239.32 g/mol
MDL Number: MFCD12198552
CAS Number: 203661-69-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR