4-Benzyloxy-1-tert-butoxycarbonylindole-2-boronic acid ≥97%

Supplier: Matrix Scientific

Synonyms: 4-Benzyloxyindole-2-boronic acid, N-BOC protected

069393-1G 069393-5G
101939-914EA 361.54 USD
101939-914 101939-916
4-Benzyloxy-1-tert-butoxycarbonylindole-2-boronic acid ≥97%
4-Benzyloxy-1-tert-butoxycarbonylindole-2-boronic acid

Hazard: Irritant

Formula: C₂₀H₂₂BNO₅
MW: 367.21 g/mol
MDL Number: MFCD04973739
CAS Number: 850568-52-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR