3-Oxo-7-azaspiro[3.5]nonane-7-carboxylate-tert-butyl ester ≥95%

Supplier: Matrix Scientific

Synonyms: tert-Butyl 1-oxo-7-azaspiro[3.5]nonane-7-carboxylate

069390-100MG 069390-250MG 069390-500MG
101939-898EA 1978.46 USD
101939-898 101939-900 101939-902
3-Oxo-7-azaspiro[3.5]nonane-7-carboxylate-tert-butyl ester ≥95%
3-Oxo-7-azaspiro[3.5]nonane-7-carboxylate-tert-butyl ester

Hazard: Irritant

Formula: C₁₃H₂₁NO₃
MW: 239.32 g/mol
MDL Number: MFCD14585368
CAS Number: 849203-60-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR