3-Aminomethyl-3-fluoropiperidine-1-carboxylic acid tert-butyl ester ≥90%

Supplier: Matrix Scientific

069387-100MG 069387-250MG 069387-500MG
101939-882EA 930.77 USD
101939-882 101939-884 101939-886
3-Aminomethyl-3-fluoropiperidine-1-carboxylic acid tert-butyl ester ≥90%
3-Aminomethyl-3-fluoropiperidine-1-carboxylic acid tert-butyl ester

Hazard: Irritant

Formula: C₁₁H₂₁FN₂O₂
MW: 232.3 g/mol
MDL Number: MFCD18375048
CAS Number: 1219832-36-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR