N-Boc-thiourea ≥95%

Supplier: Matrix Scientific

Synonyms: tert-Butyl carbamothioylcarbamate

069385-1G 069385-5G
101939-874EA 175.38 USD
101939-874 101939-876
N-Boc-thiourea ≥95%
N-Boc-thiourea

Hazard: Irritant

Formula: C₆H₁₂N₂O₂S
MW: 176.234 g/mol
MDL Number: MFCD04039987
CAS Number: 268551-65-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR