Thietane-3-carboxylic acid ≥95%

Supplier: Matrix Scientific

069384-250MG 069384-500MG 069384-1G
101939-868EA 1978.46 USD
101939-868 101939-870 101939-872
Thietane-3-carboxylic acid ≥95%
Thietane-3-carboxylic acid

Hazard: Irritant

Formula: C₄H₆O₂S
MDL Number: MFCD19229335
CAS Number: 765-55-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR