1-Aminocyclopropanemethanol ≥97%

Supplier: Matrix Scientific

Synonyms: (1-Aminocyclopropyl)methanol

069383-250MG 069383-500MG 069383-1G
101939-862EA 778.49 USD
101939-862 101939-864 101939-866
1-Aminocyclopropanemethanol ≥97%
1-Aminocyclopropanemethanol

Hazard: Irritant

Formula: C₄H₉NO
MW: 87.12 g/mol
MDL Number: MFCD01318221
CAS Number: 107017-72-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR