1,1-Diethoxy-2-iodoethane ≥95%

Supplier: Matrix Scientific

069381-1G 069381-5G
101939-854EA 815.38 USD
101939-854 101939-856
1,1-Diethoxy-2-iodoethane ≥95%
1,1-Diethoxy-2-iodoethane

Hazard: Irritant

Formula: C₆H₁₃IO₂
MDL Number: MFCD21648225
CAS Number: 51806-20-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR