Spiro[chromene-2,4'-piperidin]-4-(3H)-one

Supplier: Matrix Scientific

069377-250MG 069377-500MG
101939-850EA 1221.54 USD
101939-850 101939-852
Spiro[chromene-2,4'-piperidin]-4-(3H)-one
Spiro[chromene-2,4'-piperidin]-4-(3H)-one

Hazard: Irritant

Formula: C₁₃H₁₅NO₂
MDL Number: MFCD06738865
CAS Number: 136081-84-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR