tert-Butyl-3-oxo-1-phenyl-2,7-diazaspiro[3.5]nonane-7-carboxylate ≥96%

Supplier: Matrix Scientific

069376-100MG 069376-250MG 069376-500MG
101939-844EA 1512.31 USD
101939-844 101939-846 101939-848
tert-Butyl-3-oxo-1-phenyl-2,7-diazaspiro[3.5]nonane-7-carboxylate ≥96%
tert-Butyl-3-oxo-1-phenyl-2,7-diazaspiro[3.5]nonane-7-carboxylate

Hazard: Irritant

Formula: C₁₈H₂₄N₂O₃
MDL Number: MFCD18089860
CAS Number: 1014114-59-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR