N-Methyl-1-(3-methyloxetan-3-yl)methanamine ≥95%

Supplier: Matrix Scientific

069374-250MG 069374-500MG 069374-1G
101939-832EA 223.08 USD
101939-832 101939-834 101939-836
N-Methyl-1-(3-methyloxetan-3-yl)methanamine ≥95%
N-Methyl-1-(3-methyloxetan-3-yl)methanamine

Hazard: Irritant

Formula: C₆H₁₃NO
MDL Number: MFCD08691435
CAS Number: 915919-90-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR