3-Oxocyclobutanecarboxylic acid ≥95%

Supplier: Matrix Scientific

069370-1G 069370-5G 069370-25G
101939-826EA 15.38 USD
101939-826 101939-828 101939-830
3-Oxocyclobutanecarboxylic acid ≥95%
3-Oxocyclobutanecarboxylic acid

Hazard: Irritant

Formula: C5H6O3
MW: 114.10 g/mol
Melting Pt: 69…70 °C
MDL Number: MFCD00100900
CAS Number: 23761-23-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR