(S)-1-Boc-2-azetidinemethanol ≥95%

Supplier: Matrix Scientific

Synonyms: (S)-1-Boc-2-Azetidinemethanol, (S)-1-Boc-2-azetidinemethanol, (S)-1-(tert-Butoxycarbonyl)-2-azetidinemethanol

069369-1G 069369-5G
101939-822EA 218.46 USD
101939-822 101939-824
(S)-1-Boc-2-azetidinemethanol ≥95%
(S)-1-Boc-2-azetidinemethanol

Hazard: Irritant

Formula: C₉H₁₇NO₃
MW: 187.2362 g/mol
MDL Number: MFCD03093620
CAS Number: 161511-85-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR