N-Methylazetidin-3-amine ≥95%

Supplier: Matrix Scientific

069365-500MG 069365-1G 069365-5G
101939-800EA 801.54 USD
101939-800 101939-802 101939-804
N-Methylazetidin-3-amine ≥95%
N-Methylazetidin-3-amine

Hazard: Irritant

Formula: C₄H₁₀N₂
MDL Number: MFCD12547445
CAS Number: 247069-31-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR