3-Methoxy-3-methylazetidine ≥95%

Supplier: Matrix Scientific

069364-500MG 069364-1G 069364-5G
101939-794EA 872.31 USD
101939-794 101939-796 101939-798
3-Methoxy-3-methylazetidine ≥95%
3-Methoxy-3-methylazetidine

Hazard: Irritant

Formula: C₅H₁₁NO
MDL Number: MFCD11100996
CAS Number: 877665-31-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR