Azetidin-3-yl-N,N-dimethylmethanamine hydrochloride ≥95%
Supplier: Matrix Scientific
069361-250MG
069361-500MG
069361-1G
101939-780EA
1396.92
USD
101939-780
101939-782
101939-784
Azetidin-3-yl-N,N-dimethylmethanamine hydrochloride ≥95%
Azetidin-3-yl-N,N-dimethylmethanamine hydrochloride
Hazard: Irritant
Formula:
C₆H₁₅ClN₂ |
MDL Number:
MFCD19443908 CAS Number: 741287-55-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...