N-(Azetidin-3-yl)-N-methylacetamide hydrochloride ≥95%

Supplier: Matrix Scientific

069360-250MG 069360-500MG
101939-776EA 369.21 USD
101939-776 101939-778
N-(Azetidin-3-yl)-N-methylacetamide hydrochloride ≥95%
N-(Azetidin-3-yl)-N-methylacetamide hydrochloride

Hazard: Irritant

Formula: C₆H₁₃ClN₂O
MDL Number: MFCD10565806
CAS Number: 935668-15-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR