N,N-Dimethylazetidine-3-carboxamide hydrochloride ≥95%

Supplier: Matrix Scientific

069358-250MG 069358-500MG
101939-768EA 1280 USD
101939-768 101939-770
N,N-Dimethylazetidine-3-carboxamide hydrochloride ≥95%
N,N-Dimethylazetidine-3-carboxamide hydrochloride

Hazard: Irritant

Formula: C₆H₁₃ClN₂O
MDL Number: MFCD10565797
CAS Number: 927390-60-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR