3-Chloroazetidine hydrochloride ≥95%
Supplier: Matrix Scientific
069357-1G
069357-5G
069357-25G
101939-762EA
207.69
USD
101939-762
101939-764
101939-766
3-Chloroazetidine hydrochloride ≥95%
3-Chloroazetidine hydrochloride
Hazard: Irritant
Formula:
C₃H₇Cl₂N |
MDL Number:
MFCD09834116 CAS Number: 313468-63-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...