3-Methylazetidine hydrochloride ≥95%

Supplier: Matrix Scientific

069355-1G 069355-5G
101939-754EA 167.69 USD
101939-754 102551-746
3-Methylazetidine hydrochloride ≥95%
3-Methylazetidine hydrochloride

Hazard: Irritant

Formula: C4H10ClN
MDL Number: MFCD09743448
CAS Number: 935669-28-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR