N-Methylcyclopropanamine ≥95%

Supplier: Matrix Scientific

069354-1G 069354-5G
101939-746EA 209.23 USD
101939-746 101939-748
N-Methylcyclopropanamine ≥95%
N-Methylcyclopropanamine

Hazard: Irritant

Formula: C₄H₉N
MDL Number: MFCD00045391
CAS Number: 5163-20-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR