4,4-Difluoroadamantane-1-carboxylic acid methyl ester ≥95%

Supplier: Matrix Scientific

Synonyms: Methyl 4,4-difluoroadamantane-1-carboxylate

069352-250MG 069352-500MG
101939-736EA 1426.15 USD
101939-736 101939-738
4,4-Difluoroadamantane-1-carboxylic acid methyl ester ≥95%
4,4-Difluoroadamantane-1-carboxylic acid methyl ester

Hazard: Irritant

Formula: C₁₂H₁₆F₂O₂
MW: 230.26 g/mol
MDL Number: MFCD18800658
CAS Number: 1313739-04-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR