4-(Aminomethyl)phenylboronic acid pinacol ester hydrochloride ≥96%

Supplier: Matrix Scientific

Synonyms: 4-(Aminomethyl)benzeneboronic acid pinacol ester hydrochloride, 4-(Aminomethyl)phenylboronic acid pinacol ester hydrochloride, (4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)methanamine hydrochloride, [4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]methylamine hydrochloride, 4-Aminomethylphenylboronic acid pinacol ester HCl

069350-1G 069350-5G
101939-732EA 872.31 USD
101939-732 101939-734
4-(Aminomethyl)phenylboronic acid pinacol ester hydrochloride ≥96%
4-(Aminomethyl)phenylboronic acid pinacol ester hydrochloride

Hazard: Irritant

Formula: C₁₃H₂₀BNO₂·HCl
MDL Number: MFCD02179455
CAS Number: 850568-55-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR