4-(4-Bromophenyl)piperidine-4-carbonitrile ≥95%

Supplier: Matrix Scientific

069346-1G 069346-5G
101939-724EA 989.23 USD
101939-724 101939-726
4-(4-Bromophenyl)piperidine-4-carbonitrile ≥95%
4-(4-Bromophenyl)piperidine-4-carbonitrile

Hazard: Irritant

Formula: C₁₂H₁₃BrN₂
MDL Number: MFCD18433614
CAS Number: 1255666-69-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR