tert-Butyl-4-(2,2,2-trifluoro-1-(hydroxyimino)ethyl)piperidine-1-carboxylate ≥95%

Supplier: Matrix Scientific

069345-250MG 069345-500MG
101939-720EA 930.77 USD
101939-720 101939-722
tert-Butyl-4-(2,2,2-trifluoro-1-(hydroxyimino)ethyl)piperidine-1-carboxylate ≥95%
tert-Butyl-4-(2,2,2-trifluoro-1-(hydroxyimino)ethyl)piperidine-1-carboxylate

Hazard: Irritant

Formula: C₁₂H₁₉F₃N₂O₃
MW: 296.29 g/mol
MDL Number: MFCD17015048
CAS Number: 1313739-03-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR